Glycerol, 99+%, Certified AR for Analysis, Fisher Chemical™
Glycerol, 99+%, Certified AR for Analysis, Fisher Chemical™
Glycerol, 99+%, Certified AR for Analysis, Fisher Chemical™
Glycerol, 99+%, Certified AR for Analysis, Fisher Chemical™
C3H8O3, CAS Number-56-81-5, propanetriol, trihydroxypropane, glycyl alcohol, glycerol, glyceritol, glycerin, glycerine, osmoglyn, 1,2,3-propanetriol, 1,2,3-trihydroxypropane, 2.5L, 290 deg.C, CHEBI:17754, Undesignated, 92.09g/mol, PEDCQBHIVMGVHV-UHFFFAOYSA-N, propane-1,2,3-triol, 18 deg.C, 4722 | CAS: 56-81-5 | C3H8O3 | 92.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
500 mL
1 L
5 L
25 L
Packaging:
View all
HDPE plastic bottle
Catalog number FKCG065017
also known as FKCG/0650/17
Price (MYR)
284.00
Bottle
Quantity:
2.5 L
Packaging:
HDPE plastic bottle
Request bulk or custom format
Price (MYR)
284.00
Bottle
Specifications
CAS56-81-5
Melting Point18°C
pH5
Boiling Point290°C
PackagingHDPE plastic bottle
View more
C3H8O3, CAS Number-56-81-5, glycerol, propanetriol, glycyl alcohol, glycerin, 1,2,3-trihydroxypropane, glycerine, 1,2,3-propanetriol, trihydroxypropane, osmoglyn, glyceritol, 1L, 290 deg.C, CHEBI:17754, Undesignated, 92.09g/mol, PEDCQBHIVMGVHV-UHFFFAOYSA-N, propane-1,2,3-triol, 18 deg.C, 4722