Glycerol (Molecular Biology), Fisher BioReagents™
Glycerol (Molecular Biology), Fisher BioReagents™
Glycerol (Molecular Biology), Fisher BioReagents™
Fisher Chemical
Glycerol (Molecular Biology), Fisher BioReagents™
For low-temperature storage of enzymes and bacterial cultures.
Have Questions?
Change viewbuttonViewtableView
Quantity:
4 L
1 L
Packaging:
Glass Bottle
Catalog number FBRBP229-4
Price (MYR)
714.00
Each
Quantity:
4 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
714.00
Each
Specifications
Chemical Name or MaterialGlycerol
CAS56-81-5
ChemAlert Storage SymbolGray
Health Hazard 2WARNING!
Decomposition Information>290°C
View more
For low-temperature storage of enzymes and bacterial cultures
Glycerol is used for low-temperature storage of enzymes and bacterial cultures, purification of proteins, and as a component in electrophoresis loading buffers.