Sodium perchlorate, ACS reagent, anhydrous
Sodium perchlorate, ACS reagent, anhydrous
Sodium perchlorate, ACS reagent, anhydrous
Thermo Scientific Chemicals
Sodium perchlorate, ACS reagent, anhydrous
CAS: 7601-89-0 | ClNaO4 | 122.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
Packaging:
Plastic bottle
Catalog number FSA447425000
Price (MYR)
1,028.00
Each
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,028.00
Each
Specifications
Chemical Name or MaterialSodium perchlorate
Melting Point482.0°C
CAS7601-89-0
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
May cause fire or explosion; strong oxidizer.