Magnesium chloride hexahydrate, 99%, for biochemistry
Magnesium chloride hexahydrate, 99%, for biochemistry
Magnesium chloride hexahydrate, 99%, for biochemistry
Thermo Scientific Chemicals
Magnesium chloride hexahydrate, 99%, for biochemistry
CAS: 7791-18-6 | Cl2H12MgO6 | 203.30 g/mol
Have Questions?
Catalog NumberQuantity
FSA447155000500 g
Catalog number FSA447155000
Price (MYR)
411.00
Each
Quantity:
500 g
Request bulk or custom format
Price (MYR)
411.00
Each
Specifications
Chemical Name or MaterialMagnesium chloride hexahydrate
Melting Point117.0°C
CAS7791-18-6
Quantity500 g
Assay Percent Range98.5% min. (Complexometry)