Potassium chlorate, 99+%, ACS reagent
Potassium chlorate, 99+%, ACS reagent
Potassium chlorate, 99+%, ACS reagent
Thermo Scientific Chemicals
Potassium chlorate, 99+%, ACS reagent
CAS: 3811-04-9 | ClKO3 | 122.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
5 g
500 g
Packaging:
View all
Plastic bottle
Catalog number FSA418190025
Price (MYR)
1,045.00
Each
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,045.00
Each
Specifications
Chemical Name or MaterialPotassium chlorate
Melting Point356.0°C to 368.0°C
CAS3811-04-9
ColorWhite
Health Hazard 1GHS Signal Word: Danger
View more
Oxidation reagent
RUO – Research Use Only