Sodium, 99.8%, oiled sticks, wrapped in aluminum foil
Sodium, 99.8%, oiled sticks, wrapped in aluminum foil
Sodium, 99.8%, oiled sticks, wrapped in aluminum foil
Thermo Scientific Chemicals
Sodium, 99.8%, oiled sticks, wrapped in aluminum foil
CAS: 7440-23-5 | Na | 22.99 g/mol
Have Questions?
Catalog NumberQuantity
FSA390122500250 g
Catalog number FSA390122500
Price (MYR)
472.00
Bottle
Quantity:
250 g
Request bulk or custom format
Price (MYR)
472.00
Bottle
Specifications
Chemical Name or MaterialSodium
ColorGray
CAS7440-23-5
Melting Point98.0°C
Boiling Point883.0°C