Gallium(III) nitrate hydrate, 99.9998%, (trace metal basis)
Gallium(III) nitrate hydrate, 99.9998%, (trace metal basis)
Gallium(III) nitrate hydrate, 99.9998%, (trace metal basis)
Thermo Scientific Chemicals
Gallium(III) nitrate hydrate, 99.9998%, (trace metal basis)
Gallium(III) nitrate hydrate, 99.9998%, (trace metal basis), Quantity: 100g, Packaging: Glass bottle, CAS: 69365-72-6, White, Grade: Trace Metal Basis, Melting Point: 110.0 deg.C, Molecular Formula: GaN3O9.xH2O, Molecular Weight: 255.74, Percent Purity: 99.9998% | CAS: 69365-72-6 | GaN3O9 | 255.74 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number FSA212441000
Price (MYR)
5,255.00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
5,255.00
Each
Specifications
Chemical Name or MaterialGallium(III) nitrate hydrate
Melting Point110.0°C
CAS69365-72-6
ColorWhite
Health Hazard 1GHS Signal Word: Danger
View more
for ultra trace analysis
RUO – Research Use Only