Palladium(II) chloride, 59% Pd
Palladium(II) chloride, 59% Pd
Palladium(II) chloride, 59% Pd
Thermo Scientific Chemicals
Palladium(II) chloride, 59% Pd
CAS: 7647-10-1 | Cl2Pd | 177.32 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Packaging:
Glass bottle
Catalog number FSA195200250
Price (MYR)
35,084.00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
35,084.00
Each
Specifications
Chemical Name or MaterialPalladium(II) chloride
Melting Point678.0°C to 680.0°C
CAS7647-10-1
ColorRed to Brown
Health Hazard 1GHS Signal Word: Danger