▸
Salts and Inorganics
▸
Inorganic Salts
▸
Post-Transition Metal Salts
▸
Zinc Chloride (Cryst./Certified ACS), Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Zinc Chloride (Cryst./Certified ACS), Fisher Chemical™
Have Questions?
Catalog Number
Quantity
Packaging
FCMZ33-500
500 g
Poly Bottle
Catalog number
FCMZ33-500
Price (MYR)
885.00
Each
Add to cart
Quantity:
500 g
Packaging:
Poly Bottle
Request bulk or custom format
Price (MYR)
885.00
Each
Add to cart
Specifications
Boiling Point
732°C
Chemical Name or Material
Zinc Chloride
Melting Point
293°C
CAS
7646-85-7
ChemAlert Storage Symbol