Check Order Status
Stannous Chloride Dihydrate (Crystalline/Certified ACS), Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Stannous Chloride Dihydrate (Crystalline/Certified ACS), Fisher Chemical™
Have Questions?
Catalog Number
Quantity
Packaging
FCMT142-500
500 g
Poly Bottle
Catalog number
FCMT142-500
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Poly Bottle
Request bulk or custom format
Specifications
Boiling Point
652°C
Chemical Name or Material
Stannous Chloride Dihydrate
Melting Point
37°C
CAS
10025-69-1
ChemAlert Storage Symbol