Cadmium acetate, 98%, pure, anhydrous, Thermo Scientific Chemicals
Cadmium acetate, 98%, pure, anhydrous, Thermo Scientific Chemicals
Cadmium acetate, 98%, pure, anhydrous, Thermo Scientific Chemicals
Acros Organics
Cadmium acetate, 98%, pure, anhydrous, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
Catalog number ACR454000010
Price (MYR)
-
Quantity:
1 kg
Request bulk or custom format
Specifications
Chemical Name or MaterialCadmium acetate
Melting Point255.0°C to 273.0°C
CAS543-90-8
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Very toxic to aquatic life with long lasting effects.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
May cause cancer.