Ammonium iron(II) sulfate hexahydrate, 99%, ACS reagent
Ammonium iron(II) sulfate hexahydrate, 99%, ACS reagent
Ammonium iron(II) sulfate hexahydrate, 99%, ACS reagent
Ammonium iron(II) sulfate hexahydrate, 99%, ACS reagent
Thermo Scientific Chemicals
Ammonium iron(II) sulfate hexahydrate, 99%, ACS reagent
CAS: 7783-85-9 | FeH20N2O14S2 | 392.13 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR423721000100 gPlastic bottle
Catalog number ACR423721000
Price (MYR)
442.00
Each
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
442.00
Each
Specifications
Chemical Name or MaterialAmmonium iron(II) sulfate hexahydrate
Melting Point100.0°C
CAS7783-85-9
ColorBlue-Green
Health Hazard 1GHS Signal Word: Warning