Tin(IV) acetate, 98%, extra pure
Tin(IV) acetate, 98%, extra pure
Tin(IV) acetate, 98%, extra pure
Acros Organics
Tin(IV) acetate, 98%, extra pure
CAS: 2800-96-6 | C8H12O8Sn | 354.89 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR3800500505 gGlass bottle
Catalog number ACR380050050
Price (MYR)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialTin(IV) acetate
Melting Point231.0°C to 237.0°C
CAS2800-96-6
ColorWhite
Health Hazard 1GHS Signal Word: Warning