Cadmium chloride, ACS reagent, anhydrous
Cadmium chloride, ACS reagent, anhydrous
Cadmium chloride, ACS reagent, anhydrous
Thermo Scientific Chemicals
Cadmium chloride, ACS reagent, anhydrous
CAS: 10108-64-2 | CdCl2 | 183.31 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
Packaging:
View all
Glass bottle
Catalog number ACR315270050
Price (MYR)
475.00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
475.00
Each
Specifications
Boiling Point960.0°C
Chemical Name or MaterialCadmium chloride
Melting Point568.0°C
Name NoteAnhydrous, For analysis ACS
CAS10108-64-2