Cobalt(II) nitrate hexahydrate, 99+%, for analysis
Cobalt(II) nitrate hexahydrate, 99+%, for analysis
Cobalt(II) nitrate hexahydrate, 99+%, for analysis
Thermo Scientific Chemicals
Cobalt(II) nitrate hexahydrate, 99+%, for analysis
CAS: 10026-22-9 | CoH12N2O12 | 291.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
Packaging:
Plastic bottle
Catalog number ACR219215000
Price (MYR)
1,639.00
Bottle
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,639.00
Bottle
Specifications
Chemical Name or MaterialCobalt(II) nitrate hexahydrate
Melting Point55.0°C
Name Notep.a.
CAS10026-22-9
ColorBrown to Red
View more
For analysis
RUO – Research Use Only