Kaolin, pure
Kaolin, pure
Kaolin, pure
Thermo Scientific Chemicals
Kaolin, pure
CAS: 1332-58-7 | Al2H4O9Si2 | 258.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
2.5 kg
10 kg
Packaging:
View all
Plastic Bottle
Catalog number ACR211740010
Price (MYR)
595.00
Bottle
Quantity:
1 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Price (MYR)
595.00
Bottle
Specifications
Chemical Name or MaterialKaolin
Melting Point1750.0°C
CAS1332-58-7
ColorBeige
Physical FormPowder