Potassium chlorate, 99%, extra pure, Thermo Scientific Chemicals
Potassium chlorate, 99%, extra pure, Thermo Scientific Chemicals
Potassium chlorate, 99%, extra pure, Thermo Scientific Chemicals
Acros Organics
Potassium chlorate, 99%, extra pure, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
1 kg
Packaging:
Plastic bottle
Catalog number ACR208860050
Price (MYR)
2,395.00
Each
Quantity:
5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
2,395.00
Each
Specifications
Chemical Name or MaterialPotassium chlorate
Melting Point356.0°C to 368.0°C
CAS3811-04-9
ColorWhite
Health Hazard 1GHS Signal Word: Danger
View more
Oxidation reagent
RUO – Research Use Only