Calcium hydride, ca. 93%, extra pure, approx. 1-20 mm grain size
Calcium hydride, ca. 93%, extra pure, approx. 1-20 mm grain size
Calcium hydride, ca. 93%, extra pure, approx. 1-20 mm grain size
Acros Organics
Calcium hydride, ca. 93%, extra pure, approx. 1-20 mm grain size
CAS: 7789-78-8 | CaH2 | 42.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Plastic bottle
Catalog number ACR196795000
Price (MYR)
-
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialCalcium hydride
Melting Point675.0°C
Name Noteca. 93%
CAS7789-78-8
ColorGray
View more
Adsorbent, Reduction reagent
RUO – Research Use Only