Enterprise Services
Vendor Managed Inventory Services
Service Support Plans
Instrument Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See all Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Service Support Plans
Instrument Services
New Lab Project Services
Documents and Certificates
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Chemicals
▸
Salts and Inorganics
▸
Inorganic Salts
▸
Ammonium Salts
▸
Ammonium Molybdate Tetrahydrate, Certified AR for Analysis, Fisher Chemical™
Element Search
Structure Search
Fisher Chemicals
Ammonium Molybdate Tetrahydrate, Certified AR for Analysis, Fisher Chemical™
H24MO7N6O24.4H2O, CAS Number-12054-85-2, 500g, White, 1235.86g/mol, 190 deg.C, 167059, Plastic powder jar, 5.0 to 5.5, Solid
Have Questions?
Catalog Number
Quantity
A576053
also known as
A/5760/53
500 g
Catalog number
A576053
also known as
A/5760/53
HSN
28417090
Price (MYR)
-
Quantity:
500 g
Request bulk or custom format
Specifications
Melting Point
190°C
CAS
12054-85-2
Color
White
pH
5.0 to 5.5
Physical Form
Solid
View more
H24MO7N6O24.4H2O, CAS Number-12054-85-2, 500g, White, 1235.86g/mol, 190 deg.C, 167059, Plastic powder jar, 5.0 to 5.5, Solid