Silver nitrate, 99.85%, for analysis
Silver nitrate, 99.85%, for analysis
Silver nitrate, 99.85%, for analysis
Thermo Scientific Chemicals
Silver nitrate, 99.85%, for analysis
CAS: 7761-88-8 | AgNO3 | 169.87 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA197681000100 gGlass bottle
Catalog number FSA197681000
Price (MYR)
5,355.00
Bottle
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
5,355.00
Bottle
Specifications
Chemical Name or MaterialSilver nitrate
CAS7761-88-8
ColorWhite
Health Hazard 1Danger
Health Hazard 2GHS P Statement
Very toxic to aquatic life with long lasting effects.
Causes severe skin burns and eye damage.
May intensify fire; oxidizer.
View more
For analysis
RUO – Research Use Only