Calcium chloride, 96%, for biochemistry, anhydrous
Calcium chloride, 96%, for biochemistry, anhydrous
Calcium chloride, 96%, for biochemistry, anhydrous
Thermo Scientific Chemicals
Calcium chloride, 96%, for biochemistry, anhydrous
CAS: 10043-52-4 | CaCl2 | 110.98 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number FSA192735000
Price (MYR)
1,055.00
Each
Quantity:
500 g
Request bulk or custom format
Price (MYR)
1,055.00
Each
Specifications
Boiling Point>1600.0°C
Chemical Name or MaterialCalcium chloride
Melting Point782.0°C
CAS10043-52-4
Health Hazard 1GHS Signal Word: Warning