▸
Specialty Lab Glassware
▸
Beads and Boiling Aids
▸
Zirconia Beads
Actual product may vary
Fisherbrand
Zirconia Beads
The Fisher Global Exports zirconia beads are 0.1 mm in size. They are sold in one pound bottles.
Have Questions?
Catalog Number
Quantity
FISNC0362415
453.6 g
Catalog number
FISNC0362415
Price (MYR)
-
Quantity:
453.6 g
The Fisher Global Exports zirconia beads are 0.1 mm in size. They are sold in one pound bottles.
Density of 3.7 g/cc (50% denser than glass)
Good for spores and most tissues
Diameter: 0.1 mm
Specifications
Type
Zirconia/Silica Beads
Material
Glass
Diameter (Metric)
0.1 mm
For Use With (Equipment)
Lab Homogenizer
Quantity
453.6 g