Enterprise Services
Vendor Managed Inventory Services
Service Support Plans
Instrument Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See all Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Service Support Plans
Instrument Services
New Lab Project Services
Documents and Certificates
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Chemicals
▸
Salts and Inorganics
▸
Inorganic Salts
▸
Transition Metal Salts
▸
Silver Nitrate (Crystalline/Certified ACS), Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Silver Nitrate (Crystalline/Certified ACS), Fisher Chemical™
Have Questions?
Change view
Quantity:
25 g
500 g
Packaging:
Glass Bottle
Catalog number
FGCS181-25
Price (MYR)
1,186.00
Each
Add to cart
Quantity:
25 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
1,186.00
Each
Add to cart
Specifications
Boiling Point
444°C
Chemical Name or Material
Silver Nitrate
Melting Point
212°C
CAS
7761-88-8
ChemAlert Storage Symbol
Yellow
View more