▸
Salts and Inorganics
▸
Inorganic Salts
▸
Alkali Metal Salts
▸
Sodium Phosphate Monobasic Anhydrous (Colorless-to-White Crystals), Fisher BioReagents™
Element Search
Structure Search
Fisher Chemical
Sodium Phosphate Monobasic Anhydrous (Colorless-to-White Crystals), Fisher BioReagents™
For tissue culture and molecular biology applications
Have Questions?
Change view
Quantity:
1 kg
500 g
Packaging:
View all
Poly Bottle
Catalog number
FGCBP329-1
Price (MYR)
798.00
Each
Add to cart
Quantity:
1 kg
Packaging:
Poly Bottle
Request bulk or custom format
Price (MYR)
798.00
Each
Add to cart
Specifications
Boiling Point
203.9°C
Chemical Name or Material
Sodium Phosphate Monobasic Anhydrous
Melting Point
100°C
CAS
7558-80-7
ChemAlert Storage Symbol
Gray
View more
For tissue culture and molecular biology applications
Sodium Phosphate is commonly used in tissue culture and molecular biology applications.