▸
Salts and Inorganics
▸
Inorganic Salts
▸
Alkali Metal Salts
▸
Sodium Sulfite Anhydrous (Crystalline/Certified ACS), Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Sodium Sulfite Anhydrous (Crystalline/Certified ACS), Fisher Chemical™
Have Questions?
Catalog Number
Quantity
Packaging
FCMS430-500
500 g
Glass Bottle
Catalog number
FCMS430-500
Price (MYR)
497.00
Each
Add to cart
Quantity:
500 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
497.00
Each
Add to cart
Specifications
Chemical Name or Material
Sodium Sulfite Anhydrous
Melting Point
500°C
CAS
7757-83-7
ChemAlert Storage Symbol
Gray
Color
White
View more