▸
Biochemical Reagents
▸
Carbohydrates
▸
Sugar alcohols
▸
Glycerol (Clear, Colorless/USP/FCC), Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Glycerol (Clear, Colorless/USP/FCC), Fisher Chemical™
Have Questions?
Catalog Number
Quantity
Packaging
FCMG31-4
4 L
Glass Bottle
Catalog number
FCMG31-4
Price (MYR)
772.00
Bottle
Add to cart
Quantity:
4 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
772.00
Bottle
Add to cart
Specifications
Chemical Name or Material
Glycerol
Name Note
Clear, Colorless
CAS
56-81-5
ChemAlert Storage Symbol
Gray
Health Hazard 2
WARNING!
View more