Potassium Phosphate Dibasic (Fine White Crystalline Powder), Fisher BioReagents™
Potassium Phosphate Dibasic (Fine White Crystalline Powder), Fisher BioReagents™
Potassium Phosphate Dibasic (Fine White Crystalline Powder), Fisher BioReagents™
Fisher Chemical
Potassium Phosphate Dibasic (Fine White Crystalline Powder), Fisher BioReagents™
CAS: 11-4-7758 | Molecular Formula: HK2O4P | Molecular Weight: 174.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
Packaging:
View all
Glass Bottle
Catalog number FBRBP363-500
Price (MYR)
513.00
Each
Quantity:
500 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
513.00
Each
Specifications
Chemical Name or MaterialPotassium Phosphate Dibasic
Melting Point340°C
CAS11-4-7758
ChemAlert Storage SymbolGray
ColorWhite
View more
Potassium Phosphate Dibasic (Fine White Crystalline Powder)
This potassium salt is used in buffers for molecular biology and cell culture applications.