▸
Biochemical Reagents
▸
Carbohydrates
▸
Glycosyl compounds
▸
D-Sucrose (Molecular Biology), Fisher BioReagents
Element Search
Structure Search
Fisher Chemical
D-Sucrose (Molecular Biology), Fisher BioReagents
For the preparation of density gradients for DNA ultracentrifugation
Have Questions?
Catalog Number
Quantity
Packaging
FBRBP220-212
2.5 kg
Glass Bottle
Catalog number
FBRBP220-212
Price (MYR)
736.00
Each
Add to cart
Quantity:
2.5 kg
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
736.00
Each
Add to cart
Specifications
Alpha Vector
Sucrose
Chemical Name or Material
D-Sucrose
CAS
57-50-1
ChemAlert Storage Symbol
Gray
Health Hazard 2
CAUTION!
View more
D-Sucrose is suitable for the preparation of density gradients used in purification of proteins and nucleic acids by ultracentrifugation.