▸
Salts and Inorganics
▸
Inorganic Salts
▸
Ammonium Salts
▸
Ammonium Sulfate, Fisher BioReagents
Element Search
Structure Search
Fisher Chemical
Ammonium Sulfate, Fisher BioReagents
Have Questions?
Catalog Number
Quantity
Packaging
FBRBP212R-1
1 kg
Poly Bottle
Catalog number
FBRBP212R-1
Price (MYR)
626.00
Each
Add to cart
Quantity:
1 kg
Packaging:
Poly Bottle
Request bulk or custom format
Price (MYR)
626.00
Each
Add to cart
Specifications
Chemical Name or Material
Ammonium Sulfate
Melting Point
280°C
CAS
7783-20-2
ChemAlert Storage Symbol
Gray
Color
Colorless
View more
This enzyme-grade Ammonium Sulfate is suitable for use in the purification of proteins.