Cesium Chloride (Crystalline Powder), Fisher BioReagents
Cesium Chloride (Crystalline Powder), Fisher BioReagents
Cesium Chloride (Crystalline Powder), Fisher BioReagents
Fisher Chemical
Cesium Chloride (Crystalline Powder), Fisher BioReagents
For isolation of phage DNA, plasmid DNA, or total RNA
Have Questions?
Catalog NumberQuantityPackaging
FBRBP1595-500500 gPoly Bottle
Catalog number FBRBP1595-500
Price (MYR)
3,137.00
Each
Quantity:
500 g
Packaging:
Poly Bottle
Request bulk or custom format
Price (MYR)
3,137.00
Each
Specifications
Boiling Point1290°C
Chemical Name or MaterialCesium Chloride
Melting Point646°C
CAS7647-17-8
ChemAlert Storage SymbolGray
View more
This economical grade of Cesium Chloride can be used where low levels of metal ion impurities do not interfere. It is recommended for isolating phage DNA, plasmid DNA, or total RNA.