▸
Salts and Inorganics
▸
Inorganic Salts
▸
Transition Metal Salts
▸
Copper(II) Sulfate Pentahydrate, Certified AR for Analysis, Fisher Chemical™
Element Search
Structure Search
Fisher Chemicals
Copper(II) Sulfate Pentahydrate, Certified AR for Analysis, Fisher Chemical™
Have Questions?
Change view
Quantity:
1 kg
250 g
500 g
Catalog number
C856060
also known as
C/8560/60
HSN
28332500
Price (MYR)
-
Quantity:
1 kg
Request bulk or custom format
Specifications
Melting Point
110°C
CAS
7758-99-8
Color
Blue
pH
3.5 to 4.5
Physical Form
Solid
View more