Tin(II) chloride dihydrate, low in Mercury, suitable for AAS
CAS: 10025-69-1 | Cl2Sn·2H2O | 225.63 g/mol
Have Questions?
Catalog Number
Quantity
ACR459600500
50 g
Catalog number ACR459600500
Price (MYR)
426.00
Each
Quantity:
50 g
Request bulk or custom format
Price (MYR)
426.00
Each
Specifications
Boiling Point652.0°C
Chemical Name or MaterialTin(II) chloride dihydrate
Melting Point37.0°C to 38.0°C
CAS10025-69-1
Health Hazard 1GHS Signal Word: Danger