Potassium bromide, for spectroscopy
Potassium bromide, for spectroscopy
Potassium bromide, for spectroscopy
Acros Organics
Potassium bromide, for spectroscopy
CAS: 7758-02-3 | BrK | 119.00 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
Catalog number ACR457322500
Price (MYR)
-
Quantity:
250 g
Request bulk or custom format
Specifications
Boiling Point1435.0°C
Chemical Name or MaterialPotassium bromide
Melting Point734.0°C
CAS2-3-7758
Health Hazard 1GHS Signal Word: Warning