Magnesium sulfate heptahydrate, 98+%, ACS reagent
Magnesium sulfate heptahydrate, 98+%, ACS reagent
Magnesium sulfate heptahydrate, 98+%, ACS reagent
Thermo Scientific Chemicals
Magnesium sulfate heptahydrate, 98+%, ACS reagent
CAS: 10034-99-8 | H14MgO11S | 246.47 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
25 g
500 g
Packaging:
Plastic bottle
Catalog number ACR423900010
Price (MYR)
825.00
Each
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
825.00
Each
Specifications
Chemical Name or MaterialMagnesium sulfate heptahydrate
Melting Point770.0°C
CAS10034-99-8
ColorWhite
pH5.0 to 8.2 (5% soln. at 25°C)