Lithium aluminum hydride, 95%, tablets (ca. 0.6 g)
Lithium aluminum hydride, 95%, tablets (ca. 0.6 g)
Lithium aluminum hydride, 95%, tablets (ca. 0.6 g)
Acros Organics
Lithium aluminum hydride, 95%, tablets (ca. 0.6 g)
CAS: 16853-85-3 | AlH4Li | 37.95 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ACR304531000
Price (MYR)
-
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialLithium Aluminum hydride
Melting Point125.0°C
Name Note95%
CAS16853-85-3
ColorWhite to Gray
View more
Reduction reagent
RUO – Research Use Only