Vanadium(IV) chloride, 99+%
Vanadium(IV) chloride, 99+%
Vanadium(IV) chloride, 99+%
Acros Organics
Vanadium(IV) chloride, 99+%
CAS: 7632-51-1 | Cl4V | 192.75 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number ACR299691000
Price (MYR)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Boiling Point153.0°C
Chemical Name or MaterialVanadium(IV) chloride
Melting Point-20.5°C
Name Note99+%
CAS7632-51-1