Zinc selenide, 99.99%, (trace metal basis)
Zinc selenide, 99.99%, (trace metal basis)
Zinc selenide, 99.99%, (trace metal basis)
Acros Organics
Zinc selenide, 99.99%, (trace metal basis)
CAS: 1315-09-9 | SeZn | 144.35 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number ACR223640250
Price (MYR)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialZinc selenide
Melting Point>1100°C
Name Note99.99%
CAS1315-09-9
ColorYellow
View more
for ultra trace analysis
RUO – Research Use Only