Manganese(III) acetate dihydrate, 98%
Manganese(III) acetate dihydrate, 98%
Manganese(III) acetate dihydrate, 98%
Manganese(III) acetate dihydrate, 98%
Manganese(III) acetate dihydrate, 98%
Acros Organics
Manganese(III) acetate dihydrate, 98%
CAS: 19513-05-4 | C6H9MnO6 | 232.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number ACR201740250
Price (MYR)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialManganese(III) acetate dihydrate
Name Note98%
CAS19513-05-4
ColorBrown to Brown-Orange
Health Hazard 1GHS Signal Word: Warning