Iron(II) sulfate heptahydrate, 99.5%, for analysis
Iron(II) sulfate heptahydrate, 99.5%, for analysis
Iron(II) sulfate heptahydrate, 99.5%, for analysis
Thermo Scientific Chemicals
Iron(II) sulfate heptahydrate, 99.5%, for analysis
CAS: 7782-63-0 | FeH14O11S | 278.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic bottle
Catalog number ACR201392500
Price (MYR)
561.00
Bottle
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
561.00
Bottle
Specifications
Chemical Name or MaterialIron(II) sulfate heptahydrate
Melting Point64.0°C
CAS7782-63-0
ColorBlue to Green
Health Hazard 1GHS Signal Word: Warning