Rhodium on alumina, 5% Rh, powder
Rhodium on alumina, 5% Rh, powder
Rhodium on alumina, 5% Rh, powder
Acros Organics
Rhodium on alumina, 5% Rh, powder
CAS: 7440-16-6 | Rh | 102.91 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Catalog number ACR199570050
Price (MYR)
-
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or MaterialRhodium on alumina
CAS7440-16-6
Melting Point2072.0°C
Boiling Point3147.0°C
CAS1344-28-1