Gadolinium(III) nitrate pentahydrate, 99.9%
Gadolinium(III) nitrate pentahydrate, 99.9%
Gadolinium(III) nitrate pentahydrate, 99.9%
Acros Organics
Gadolinium(III) nitrate pentahydrate, 99.9%
CAS: 52788-53-1 | GdN3O9·5H2O | 433.33 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR19914025025 gGlass bottle
Catalog number ACR199140250
Price (MYR)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialGadolinium(III) nitrate pentahydrate
Melting Point92.0°C
Name Note99.9%
CAS52788-53-1