▸
Salts and Inorganics
▸
Inorganic Salts
▸
Alkali Metal Salts
▸
Lithium hydride, 97%, pure, powder (<100 μm), Thermo Scientific™
Element Search
Structure Search
Acros Organics
Lithium hydride, 97%, pure, powder (<100 μm), Thermo Scientific™
Have Questions?
Change view
Quantity:
100 g
Catalog number
ACR191191000
Price (MYR)
-
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or Material
Lithium hydride
Melting Point
680.0°C
Name Note
98%
CAS
7580-67-8
Color
White to Gray
View more
Reduction reagent
RUO – Research Use Only