Lithium aluminum hydride, 95%, powder
Lithium aluminum hydride, 95%, powder
Lithium aluminum hydride, 95%, powder
Acros Organics
Lithium aluminum hydride, 95%, powder
CAS: 16853-85-3 | AlH4Li | 37.95 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
10 g
25 g
Catalog number ACR190325000
Price (MYR)
-
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialLithium Aluminum hydride
Melting Point125.0°C
CAS16853-85-3
Health Hazard 1Danger
Health Hazard 2GHS H Statement:
Causes severe skin burns and eye damage.
In contact with water releases flammable gases which may ignite spontaneously.
View more
Reduction reagent
RUO – Research Use Only