Sodium hydride, 60% dispersion in mineral oil, in soluble bags
Sodium hydride, 60% dispersion in mineral oil, in soluble bags
Sodium hydride, 60% dispersion in mineral oil, in soluble bags
Acros Organics
Sodium hydride, 60% dispersion in mineral oil, in soluble bags
CAS: 7646-69-7 | HNa | 24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
100 g
250 g
Catalog number ACR189860010
Price (MYR)
-
Quantity:
1 kg
Request bulk or custom format
Specifications
Chemical Name or MaterialSodium hydride, 60% dispersion in mineral oil
Melting Point800.0°C
CAS7646-69-7
ColorGray to Yellow
Health Hazard 1Danger
View more
Adsorbent, Reduction reagent
RUO – Research Use Only