Rhodizonic acid, disodium salt, 98%
Rhodizonic acid, disodium salt, 98%
Rhodizonic acid, disodium salt, 98%
Rhodizonic acid, disodium salt, 98%
Acros Organics
Rhodizonic acid, disodium salt, 98%
CAS: 523-21-7 | C6Na2O6 | 214.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
Catalog number ACR132341000
Price (MYR)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialRhodizonic acid, disodium salt
Melting Point>300.0°C
CAS523-21-7
ColorGray-Green to Green
Physical FormFine Crystalline Powder
View more
Indicator
RUO – Research Use Only