RO-TAP™ Sieve Shaker Sound Enclosure
Actual product may vary
W.S. TYLER
RO-TAP™ Sieve Shaker Sound Enclosure
For RO-TAP sieve shakers
Have Questions?
Catalog NumberFor Use With (Equipment)
TYLR-30050Ro-Tap sieve shakers
Catalog number TYLR-30050
Price (MYR)
11,606.00
Each
For Use With (Equipment):
Ro-Tap sieve shakers
Price (MYR)
11,606.00
Each
For RO-TAP sieve shakers
  • Heavy 18-gauge steel construction
  • Fully insulated with 1 in. thick (2.5cm) metal-faced acoustic foam inner lining effectively reduces noise levels from approximately 85 to 60dB
  • Hinged top cover with automatic lift cylinders, latches, and air ventilation hose
  • Gray textured finish
Specifications
Product TypeSieve Shaker Sound Enclosure
Material18 G Steel, Metal-faced foam liner
Diameter (English)24 in.
Diameter (Metric)60.96 cm
Dimensions (L x W x H)24 x 35 x 29 in. (610 x 88.9 x 73.66 cm)
Height (English)29 in.
Height (Metric)73.66 cm
Length (English)24 in.
Length (Metric)610 mm
Thickness (Metric)2.5 cm
Width (English)35 in.
Width (Metric)88.9 cm
Weight (English)90 lb.
Weight (Metric)40.8 kg
For Use With (Equipment)Ro-Tap sieve shakers
Unit SizeEach