Arachidonic Acid Reagent
Arachidonic Acid Reagent
Actual product may vary
W.S. TYLER
Arachidonic Acid Reagent
Arachidonic Acid Reagent is used in platelet aggregation studies for abnormal function induced by aspirin and aspirin-containing medications
Have Questions?
Catalog NumberQuantity
TYL53642 x 1 mL
Catalog number TYL5364
Price (MYR)
956.00
EA
Add to cart
Quantity:
2 x 1 mL
Price (MYR)
956.00
EA
Add to cart
Arachidonic Acid Reagent is used in platelet aggregation studies for abnormal function induced by aspirin and aspirin-containing medications
  • Reagents are optimized for use with AggRAM, but may be used on any turbidometric aggregometer of similar quality
  • Arachidonic Acid is a lyophilized preparation of sodium arachidonate for use in platelet aggregation studies of abnormal function, especially dysfunction induced by aspirin and aspirin-containing medications
  • The concentration of the reconstituted reagent is 5 mg/mL
Specifications
Concentration5 mg/mL
Detectable AnalytesPlatelets
For Use With (Equipment)AggRAM
Sample TypePlasma
Quantity2 x 1 mL
FormLyophilized
Unit SizeEA
Contents & Storage
2°C to 8°C