Nancy B's Science Club™ Way to Grow™ Hydroponics Kit and Gardening Diary
Nancy B's Science Club™ Way to Grow™ Hydroponics Kit and Gardening Diary
Actual product may vary
W.S. TYLER
Nancy B's Science Club™ Way to Grow™ Hydroponics Kit and Gardening Diary
Sprout seeds, put them in the seed baskets, place seed baskets in the water-filled flasks, and watch your plants climb the trellis as they grow. Set includes everything kids need to explore the world of hydroponics, including 2 seed baskets, 2 flasks with thermometers, and 2 trellises with rulers
Have Questions?
Catalog number TYL5361
View Price:Sign in to see your account pricing. Need an account? Register with us today.
  • Sprout seeds, put them in the seed baskets, place seed baskets in the water-filled flasks, and watch your plants climb the trellis as they grow
  • This set includes everything kids need to explore the amazing world of hydroponics, including 2 seed baskets, 2 flasks with thermometers, and 2 trellises with rulers.
  • And the included, 22-page journal is packed with experiments, fun facts, creative writing prompts, and more
  • Stimulate an interest in biology and botany with this Nancy B's Science Club set
  • Perfect for science fair projects and STEM learning.
Specifications
Product TypeGardening Kit
GradeK to 12
Class Size2 Students
Unit SizeEA