Depyrogenated Sterile Empty Vials
Actual product may vary
Depyrogenated Sterile Empty Vials
Depyrogenated Sterile Empty Vials
Depyrogenated Sterile Empty Vials
Depyrogenated Sterile Empty Vials
Depyrogenated Sterile Empty Vials
Depyrogenated Sterile Empty Vials
Thermo Scientific
Depyrogenated Sterile Empty Vials
Thermo Scientific™ Sterile Empty Vials are offered in capacities ranging from 1 to 100mL.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)ColorClosure SizeNo. per Case
ST10-2010 mLClear20 mm50
FIS12-200-1482 mLClear13 mm100
FIS12-200-15030 mLClear20 mm50
FIS12-200-15150 mLClear20 mm50
FIS12-200-145100 mLClear20 mm50
Catalog number ST10-20
HSN 70179090
Price (MYR)
-
Volume (Metric):
10 mL
Color:
Clear
Closure Size:
20 mm
No. per Case:
50
Thermo Scientific™ Depyrogenated Sterile Empty Vials are offered in capacities ranging from 1 to 100mL. Certificate of Sterility & Pyrogen Test included with each lot. Sterile vials are Type I borosilicate, assembled with butyl stoppers and aluminum seals.
  • Type 1 Borosilicate glass
  • Assembled wiht butyl stoppers and aluminum seals
  • Suitable for uses where aseptic protocol is required
  • Certificate of Sterility & Pyrogen Test included with each lot
Specifications
DescriptionClear glass vial pre-assembled with stopper and aluminum crimp seal
Diameter (English) Outer0.813 in.
ColorClear
Diameter (Metric) Outer20.65 mm
MaterialGlass
SterilitySterile
Volume (Metric)10 mL
Height (English)2 in.
No. per Case50
Volume (English)0.34 oz.
Volume (Metric) Total10 mL
Closure MaterialAluminum
Closure TypeCrimp Cap (Assembled) with Stopper
Quantity50 Cs.
Closure Size20 mm
For Use With (Equipment)Aseptic Protocol
Certifications/ComplianceCertificates of Sterility and Pyrogen Test
Height (Metric)50.8 mm
Product TypeVial
Unit SizeBox