▸
Chromatography Supplies
▸
Chromatography Port Injection Liners
▸
GC Inlet Liner Split/Splitless with Single Taper Quartz Wool For Agilent Ac Control Injector 4mm I.D.
Actual product may vary
GC Inlet Liner Split/Splitless with Single Taper Quartz Wool For Agilent Ac Control Injector 4mm I.D.
The unique deactivation process of SGE inlet liners provides excellent analysis for a wide range of compounds
Have Questions?
Catalog Number
Quantity
SGE092218
25/Pk.
Catalog number
SGE092218
Price (MYR)
3,322.00
Pack
Add to cart
Quantity:
25/Pk.
Price (MYR)
3,322.00
Pack
Add to cart
Each batch of liners tested for activity
Manufactured from borosilicate glass
Color coded by geometry for easy identification
For use with Thermo Scientific instruments
Specifications
Quantity
25/Pk.
Length (Metric)
78.5 mm
Product Type
GC Inlet Liner, Split/Splitless with Single Taper (Quartz Wool)
Diameter (Metric) Inner
4 mm
Diameter (Metric) Outer
6.3 mm